missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Microflex™ 93-260 Chemical-Resistant Disposable Glove
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Three layer design for superior protection against harsh chemicals including acids, solvents and bases
- Thin mil construction provides enhanced tactility and dexterity
- Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
- Lower acceptable pinhole rate (0.65 AQL) and extended cuff for reliable protection against hazardous substances
- Silicone free formulation and processing ensure better product protection
SIZE CHART
- XS – 5.5 to 6
- S – 6.5 to 7
- M – 7.5 to 8
- L – 6.5 to 9
- XL – 9.5 to 10
- 2XL – 10.5 to 11

Specifications
Specifications
Certifications/Compliance | ASTM D3577, TYPE II, PPE 89/686, EN 420, 388 & 374 CATEGORY III |
Color | Green, Blue |
Disposable | Single-use |
Hand | Ambidextrous |
Material | Nitrile, Neoprene |
Powdered | Powder Free |
Product Type | Disposable Gloves |
Size | Small |
Thickness (Metric) | 0.198 mm |
Finger Style | Textured |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 14
Videos
-
Ansell - Microflex 93-260 Gloves
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
22Hours
22Minutes
48Seconds
44 -
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell™ Microflex™ 93-260 Chemical-Resistant Disposable Glove >
Spot an opportunity for improvement?Share a Content Correction