Learn More
Citric acid monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 135.0°C to 152.0°C |
Flash Point | 174°C |
Ignition Residue | 0.02% max |
Assay Percent Range | 99+% |
Packaging | Plastic bottle |
Linear Formula | HO2CCH2C(OH)(CO2H)CH2CO2H·H2O |
Quantity | 1 kg |
Merck Index | 15,2325 |
Solubility Information | Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol |
Formula Weight | 210.15 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468217 | Certificate of Analysis | 31/01/2025 | 385850010,385850025,385852500 |
A0462421 | Certificate of Analysis | 01/05/2024 | 385850010,385850025,385852500 |
A0450691 | Certificate of Analysis | 02/05/2023 | 385850010,385850025,385852500 |
A0402823 | Certificate of Analysis | 13/12/2022 | 385850010,385850025,385852500 |
A0412546 | Certificate of Analysis | 13/12/2022 | 385850010,385850025,385852500 |
Safety and Handling

- Citric acid monohydrate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
0Hours
19Minutes
6Seconds
41
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.