missing translation for 'onlineSavingsMsg'
Learn More
Citric acid monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 12095350

Citric acid monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 12095350
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 12095350

Brand: Thermo Scientific Acros 385852500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5949-29-1
Molecular Formula C6H8O7·H2O
Molecular Weight (g/mol) 210.15
InChI Key YASYEJJMZJALEJ-UHFFFAOYSA-N
Synonym citric acid monohydrate, citric acid hydrate, 2-hydroxypropane-1,2,3-tricarboxylic acid hydrate, citric acid, monohydrate, unii-2968phw8qp, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, monohydrate, citrate, acidum citricum monohydricum, citric acid monohydrate uspShow MoreShow Less
PubChem CID 22230
ChEBI CHEBI:31404
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;hydrate
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O

Specifications

Melting Point 135.0°C to 152.0°C
Flash Point 174°C
Ignition Residue 0.02% max
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula HO2CCH2C(OH)(CO2H)CH2CO2H·H2O
Quantity 250 g
Merck Index 15,2325
Solubility Information Solubility in water: 676g/L (25°C). Other solubilities: 2.17g/100g ether - 0.007g/100g chloroform, 15.43g/100g amyl alcohol - 5.98g/100g amyl acetate, 5.28g/100g ethyl acetate - 197g/100g methanol, 62.8g/100g propanol
Formula Weight 210.15
Percent Purity 99+%
Grade ACS Reagent
Insoluble Matter 0.005% max
Chemical Name or Material Citric acid monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468217 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number385850010,385850025,385852500
Lot NumberA0462421 Certificate TypeCertificate of Analysis Date01/05/2024 Catalog Number385850010,385850025,385852500
Lot NumberA0450691 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number385850010,385850025,385852500
Lot NumberA0402823 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number385850010,385850025,385852500
Lot NumberA0412546 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number385850010,385850025,385852500
compliance-icons
Product Identifier
  • Citric acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citric acid monohydrate, 99+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.