missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Comfort Rack for Upright Freezers, 75 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand: Tenak™ TE24276
This item is not returnable.
View return policy
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
Specifications
75 mm (Box) | |
Upright Freezers | |
4 x 4 |
562 x 140 x 328 mm | |
Boxes (16) | |
Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tenak™ Comfort Rack for Upright Freezers, 75 mm Box Height > Holds: 16 Boxes (4 x 4)
Spot an opportunity for improvement?Share a Content Correction