missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Conical Centrifuge Tubes

Ideal for applications requiring larger sample volumes.

Brand:  Thermo Scientific™ 332265

133.24 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 17554725

  • £166.00 / Pack of 48
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ Conical Centrifuge Tubes are sterile and ideal for applications requiring larger sample volumes.

  • Manufactured with high quality virgin resins
  • Sterilized by gamma irradiation per ISO 11137-2
  • Non-pyrogenic
  • Non-cytotoxic
  • DNase/RNase free
  • Human DNA free
  • BSE/TSE free

Compliance

Compliant with California Prop 65

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Transparent
Single-use
None
Sterile
Conical
8 oz.
No
Yes
7,000 x g
250 mL
Polypropylene
6
Not Autoclavable
162 mm
Conical
60 mm
Centrifuge Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Conical Centrifuge Tubes >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.