missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CTS™ AIM V™ SFM
GREENER_CHOICE

Gibco CTS AIM V SFM (Therapeutic Grade) is the first commercially available, defined, serum-free medium (SFM) for the proliferation and/or manipulation of T cells and dendritic cells manufactured in compliance with cGMP.

Brand:  Gibco™ 0870112DK

 View more versions of this product

Product Code. 11534536

  • £185.00 / 1000mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Gibco CTS AIM V SFM (Therapeutic Grade) is the first commercially available, defined, serum-free medium (SFM) for the proliferation and/or manipulation of T cells and dendritic cells manufactured in compliance with cGMP. It contains L-glutamine, 50 μg/mL streptomycin sulfate, and 10 μg/mL gentamicin sulfate.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
Liquid
Serum-free
Cell and Gene Therapy Research, Development, and Manufacturing
1 L
Bottle
Animal Origin
Cell Culture Medium
Sterile-filtered
ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Ambient
Store at 2–8°C and protect from light.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2445790 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number0870112BK,0870112DK
Lot Number2495860 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number0870112BK,0870112DK
Lot Number2561513 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number0870112BK,0870112DK
Lot Number2337577 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number0870112BK,0870112DK
Lot Number2462025 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number0870112BK,0870112DK
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ CTS™ AIM V™ SFM > Quantity: 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.