missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Cytidine-5'-diphosphocholine sodium salt Product Code.: 15466239

Thermo Scientific Chemicals Cytidine-5'-diphosphocholine sodium salt

Product Code. 15466239
25 g
Click to view available options
Quantity:
10 g
25 g
50 g
This item is not returnable. View return policy

Product Code. 15466239

Brand: Thermo Scientific Chemicals J64161.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Intermediate in the synthesis of phosphatidylcholine; also a phospholipase inhibitor

Cytidine-5'-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cytidine-5′-diphosphocholine sodium salt hydrate acts as a neuroprotective agent in situations of hypoxia and ischemia. It is used as biosynthetic intermediate of membrane phospholipids. It is a cerebral vasodilator and inhibits PLA2 activation.

Solubility
Soluble in water (100 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33818-15-4
Molecular Formula C14H25N4NaO11P2
Molecular Weight (g/mol) 510.31
MDL Number MFCD00150508
InChI Key YWAFNFGRBBBSPD-OCMLZEEQSA-M
Synonym citicolinesodium, cytidine 5'-diphosphocholine sodium, cdp-choline sodium, citicoline, sodium, cdp-choline, sodium, cytidine 5'-diphosphocholine sodium salt, cytidine 5'-diphosphocholine sodium salt dihydrate, cytidine-5'-diphosphocholine monosodium salt, sodium 2s,3r,4s,5s-5-4-amino-2-oxo-1-pyrimidinyl-3,4-dihydroxy-2-oxolanyl methoxy-oxidophosphoryl 2-trimethylammonio ethyl phosphate, sodium 2s,3r,4s,5s-5-4-azanyl-2-oxidanylidene-pyrimidin-1-yl-3,4-bis oxidanyl oxolan-2-yl methoxy-oxidanidyl-phosphoryl 2-trimethylazaniumyl ethyl phosphateShow MoreShow Less
PubChem CID 46738085
IUPAC Name sodium;[[(2S,3R,4S,5S)-5-(4-amino-2-oxopyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] 2-(trimethylazaniumyl)ethyl phosphateShow MoreShow Less
SMILES [Na+].C[N+](C)(C)CCOP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(N)=NC1=O

Specifications

Color White
Quantity 25 g
Beilstein 4110040
Solubility Information Soluble in water (100mg/ml).
Formula Weight 510.31 (Anhydrous)
Physical Form Powder
Chemical Name or Material Cytidine-5'-diphosphocholine sodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS28I019 Certificate TypeCertificate of Analysis Date07/07/2022 Catalog NumberJ64161.14,J64161.18,J64161.09
Lot NumberP24I034 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberJ64161.14,J64161.18,J64161.09
Lot NumberU21D043 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ64161.14,J64161.18,J64161.09
Lot NumberM24D035 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ64161.14,J64161.18,J64161.09
Lot NumberY16D068 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ64161.14,J64161.18,J64161.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Cytidine-5'-diphosphocholine sodium salt >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.