missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Dispensette™ S Analog-adjustable Bottletop Dispensers
GREENER_CHOICE

The Dispensette™ S Analog Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.

Brand:  BRAND™ 4600161

491.09 GBP valid until 2025-03-30
Use promo code "24134" to get your promotional price.


 View more versions of this product

Product Code. 15406932

  • ÂŁ715.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual

Description

Description

  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Safe volume setting with internal scalloped track (analog-adjustable instruments)
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking

Services

One year

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5% (250ÎĽL)
Autoclavable (121°C)
Red
5 to 50 mL
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
Bottletop Dispenser
ISO 9001
With recirculation valve
Analog
1.0 mL
Product Suggestions

Product Suggestions

Videos

Videos

  • Brand Dispensette S Accessories
    Brand Dispensette S Accessories
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Brand Dispensette S Accessories

    • Brand Dispensette S Cleaning
      Brand Dispensette S Cleaning
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Brand Dispensette S Cleaning

      • Brand Dispensette S Teaser
        Brand Dispensette S Teaser
        Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
         
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected

          Brand Dispensette S Teaser

        • Brand Dispensette S Startup
          Brand Dispensette S Startup
          Video Player is loading.
          Current Time 0:00
          Duration 0:00
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 0:00
           
          1x
          • Chapters
          • descriptions off, selected
          • captions off, selected

            Brand Dispensette S Startup

          SDS
          Documents Certificates
          Special Offers

          Special Offers

          Product Content Correction

          Your input is important to us. Please complete this form to provide feedback related to the content on this product.

          Product Title
          BRAND™ Dispensette™ S Analog-adjustable Bottletop Dispensers > With recirculation valve; Volume: 5-50mL

          By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

          Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.