missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Dispensette™ S Digital Bottletop Dispensers
GREENER_CHOICE

The Dispensette™ S Digital Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.

Brand:  BRAND™ 4600321

409.79 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.


 View more versions of this product

Product Code. 15428073

  • ÂŁ560.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual
This item is not returnable. View return policy

Description

Description

The Dispensette™ S Digital Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for g

  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Easy Calibration technique for calibration adjustment in seconds without tools
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replacable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking

Services

One year

TRUSTED_SUSTAINABILITY
Specifications

Specifications

0.5% (10ÎĽL)
Autoclavable (121°C)
Red
0.2 to 2 mL
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
Dispenser
Certifications: ISO 9001
With recirculation valve
Digital
0.01 mL
Product Suggestions

Product Suggestions

Videos

Videos

  • Brand Dispensette S Accessories
    Brand Dispensette S Accessories
    Video Player is loading.
    Current Time 0:00
    Duration 0:52
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:52
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Brand Dispensette S Accessories

  • Brand Dispensette S Cleaning
    Brand Dispensette S Cleaning
    Video Player is loading.
    Current Time 0:00
    Duration 4:33
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 4:33
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Brand Dispensette S Cleaning

  • Brand Dispensette S Teaser
    Brand Dispensette S Teaser
    Video Player is loading.
    Current Time 0:00
    Duration 0:34
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:34
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Brand Dispensette S Teaser

  • Brand Dispensette S Startup
    Brand Dispensette S Startup
    Video Player is loading.
    Current Time 0:00
    Duration 2:19
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 2:19
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Brand Dispensette S Startup

SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Dispensette™ S Digital Bottletop Dispensers > With recirculation valve; Volume: 0.2-2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.