missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ ECO AluRack for Chest Freezers, 50 mm Box Height
ECO AluRack fits into any chest freezer and can be used with all standard cryoboxes.
Brand: Tenak™ TE11512
This item is not returnable.
View return policy
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without standard white cardboard boxes
Specifications
Seawater resistant | |
Aluminum | |
Chest Frseezers | |
50 mm (Box) |
Boxes | |
10 | |
140 x 140 x 555 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tenak™ ECO AluRack for Chest Freezers, 50 mm Box Height > Holds: 10 Boxes
Spot an opportunity for improvement?Share a Content Correction