Learn More
Fisherbrand™ Disposable Inoculating Loops
These Disposable Inoculating Loops are smooth and flexible to facilitate smooth, uniform streaking without damaging gel surfaces. Sterile and non-sterile options available.
Brand:  Fisherbrand™ 01189165
 View more versions of this product
Product Code. 17783667
  • £146.64 / Pack of 250
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for anaerobic microbiology applications where deep agar inoculation is required
  • Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
  • High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
  • Rigid enough to penetrate the media for subsurface inoculation
  • Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
  • Can be used under hoods without danger
  • Color-coded for ease of size identification
Specifications
Specifications
Single-use
Polystyrene
Blue
Not Autoclavable
1 μL
Sterile
Cs of 250 (5 Pks. of 50/ Individually Packed)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Disposable Inoculating Loops > Disposable Inoculating Loops; Sterility: Sterile; Volume (Metric) 1.0 μL; Packaging Quantity: 50 Loops (Individually Packed)