Learn More
DWK Life Sciences DURAN™ YOUTILITY™ Pouring Ring, Cyan PP, GL 45
Shop All DWK Life Sciences ProductsDescription
The optimized, ergonomic shape of the screw cap has comfortable grooves and ridges for a more efficient and easier tightening or removal, especially with smaller sized or gloved hands. The inverted cone form of the cap makes bottle handling easier and far safer. The shortened thread inside the cap permits faster opening and closing, yet it is fully compatible with standard DIN GL 45 glass bottle threads. The tried and tested cap plug sealing system ensures a liquid tight seal. A pre-defined labelling area on the cap is compatible with the dedicated DURAN™ self-adhesive YOUTILITY™ label.
- Fully compatible with all the GL 45 threads
- The optimized, ergonomic shape of the screw cap has comfortable grooves and ridges for a more efficient and easier tightening or removal, especially with smaller sized or gloved hands
- The tried and tested cap plug sealing with sealing lip system ensures a liquid tight seal
- The pre-defined labelling areas are compatible with the dedicated DURAN™ self-adhesive YOUTILITY™ bottle labels for unambiguous bottle identification Fully compatible with all the GL 45 threads and therefore highly versatile
- Fully compatible with all the GL 45 threads and therefore highly versatile

Specifications
Specifications
Color | Light Blue |
Description | Pouring ring |
For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Storage of media and reagents; Transport and shipping |
Material | Polypropylene |
Quantity | 16/Pk. |
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
Diameter (Metric) | 53 mm |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.