Learn More
Ertugliflozin, MedChemExpress
MedChemExpress Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Brand:  Medchem Express HY-15461
Encompass_Preferred
 View more versions of this product
Product Code. 16496538
  • £84.70 / 10mg
Explore more special offers
Description
Description
Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Specifications
Specifications
Ertugliflozin
White
C22H25ClO7
PF-04971729
ClC1=CC=C([C@]23O[C@@](CO)(CO3)[C@@H](O)[C@H](O)[C@H]2O)C=C1CC4=CC=C(OCC)C=C4
436.88
Research
Metabolism-sugar/lipid metabolism
1210344-57-2
Research
10 mg
DMSO : 250 mg/mL (572.24 mM; Need ultrasonic)
436.88
98.82%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ertugliflozin, MedChemExpress