missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastDigest BstXI

Cut at CCANNNNN^NTGG sites with Thermo Scientific™ FastDigest™ BstXI, which performs best at 55°C in 5-15 minutes using universal FastDigest Buffer.

Brand:  Thermo Scientific™ FD1024

41.76 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10110740

  • £58.00 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• FastDigest BstXI (100 μL)
• 10X FastDigest Buffer (1.0 mL)
• 10X FastDigest Green Buffer (1.0 mL)

Store at –20°C.

BstXI
Yes
No
Restriction Enzyme
FastDigest™
80°C
Dcm Methylation-Sensitive, Not Dam Methylation-Sensitive, Not CpG Methylation-Sensitive
10X FastDigest Buffer/FastDigest Green Buffer
37°C
100 μL (100 Reactions)
Traditional Cloning
3' protruding ends
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2861208 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberFD1024
Lot Number2789256 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberFD1024
Lot Number2828663 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberFD1024
Lot Number2810524 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberFD1024
Lot Number2795794 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberFD1024
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ FastDigest BstXI > 100μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.