Learn More
Thermo Scientific™ Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits

Enjoy the convenience of having a variety of Finnpipette™ F2 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab
Brand: Thermo Scientific™ 4701070
602.94 GBP valid until 2025-12-31
Use promo code "24294" to get your promotional price.
Description
Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F2 GLP (Good Laboratory Pipetting) Kits provide durable, high-performance Finnpipette F2 pipettes which offer maximum reliability and comfort. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling.
Features:
- AVG-volume gearing mechanism for accuracy and precision
- Fully autoclavable for protection
- Soft-touch tip ejector for light tip ejection
- Large display for better vision ergonomics
- Very light pipetting forces
Kit 1 includes:
- Three Thermo Scientific Finnpipette Pipetters from 1 to 1000 μL:
- Finnpipette F2
- 1-10 μL
- 10-100 μL
- 100-1000 μL
- Flex tips: 10, 1 x 96; 200, 1 x 96; 1000, 1 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 2 includes:
- Four Thermo Scientific Finnpipette Pipetters from 0.2 to 1000 μL:
- Finnpipette F2
- 0.2-2 μL
- 2-20 μL univ
- 20-200 μL
- 100-1000 μL
- Flex tips: 10, 12 x 96; 200, 2 x 96; 1000, 1 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 3 includes:
- Three Thermo Scientific Finnpipette Pipetters from 10 to 10,000 μL:
- Finnpipette F2
- 10-100 μL, 100-1000 μL
- 1000-10,000μL
- Flex tips: 200, 1 x 96; 1000, 1 x 96; FT 10mL, 1 x 24 F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Kit 4 includes:
- Three Thermo Scientific Finnpipette Pipetters from 20 to 1000μL:
- Finnpipette F2
- 2-20μL, 20-200μL
- 100-1000 μL
- Flex tips: (2) 200 μL, 10 x 96; 100-1000 μL, 10 x 96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack
Ordering Information:
The kit contains samples of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains all the information of pipetting from choosing the right type to detailed de-contamination information.
Warranty: 5 years with web registration.

Specifications
Flex Tips: 200, 1000 μL | |
Autoclavable | |
F2 | |
GLP Kit |
Finnpipette F2 GLP Kit 4: 2-20μL, 20-200μL and 100-1000μL | |
Assorted | |
Finnpipette F2 | |
2 to 1000 μL |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
8Hours
14Minutes
9Seconds
51 -
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
25Hours
14Minutes
9Seconds
51
Your input is important to us. Please complete this form to provide feedback related to the content on this product.