Learn More
Thermo Scientific™ Finnpipette™ F2 Multichannel Pipettes
GREENER_CHOICE
Use this multichannel pipette for highly demanding applications where robustness and durability are essential.
Brand:  Thermo Scientific™ 4662060
585.77 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
 View more versions of this product
Product Code. 11867381
  • £1525.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Show More
Specifications
Specifications
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, 250 Wide
Autoclavable
12
Pipette
0.2μL
Finnpipette™ F2
Yellow
Finnpipette™ F2
10 to 100 μL
Yellow
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Finnpipette™ F2 Multichannel Pipettes > Finnpipette™ F2 12-Channel Pipette, Volume range: 10 to 100μL; Color Code: Yellow; Compatible tips: Finntip 250, Filter 100; Flex 200, Flex Filter 100, 200 Ext and Filter 100 Ext; Unit Size: 1 piece