missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Polypropylene Disposable Beakers
Click to view available options
Capacity (Metric):
15 mL
30 mL
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
Description
Rib construction permits stacking without binding
- Special “dripless” lip
- Base allows beaker to sit flat during use
- Graduated in mL and oz.
Specifications
Specifications
Capacity (Metric) | 100 mL |
Material | Polypropylene |
For Use With (Application) | Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents |
Includes | Spout |
Increments | 5 mL |
Type | Beaker |
Autoclavable | Autoclavable |
Disposable | Single-use |
Graduated | Yes |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Documents
Scientific Resources
Brochures
Fisherbrand_Disposable_Beaker_EN.pdf
File size: 139.8 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ Polypropylene Disposable Beakers >
Spot an opportunity for improvement?Share a Content Correction