Learn More
Gibco™ DMEM/F-12

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells.
Brand: Gibco™ 11320033
19.64 GBP valid until 2025-12-31
Use promo code "24273" to get your promotional price.
Description
DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM/F-12 is modified as follows:
With: L-glutamine, Phenol Red
Without: HEPES
The complete formulation is available.
Using DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with F-12's wide variety of components. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Order Info
Shipping Condition: Room Temperature

Specifications
MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts | |
Animal Origin-free | |
Liquid | |
Standard Serum Supplementation | |
Sterile-filtered | |
No HEPES | |
Gibco™ | |
Room Temperature |
Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells | |
1 X | |
DMEM (Dulbecco's Modified Eagle Medium)/F-12 | |
Sterile-filtered | |
High Glucose, Glutamine, Phenol Red, Sodium Pyruvate | |
cGMP-compliant under the ISO 13485 standard | |
500 mL | |
Storage conditions: 2°C to 8°C (protect from light) Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2661508 | Certificate of Analysis | 31/01/2025 | 11320033 |
2645225 | Certificate of Analysis | 29/01/2025 | 11320033 |
2645233 | Certificate of Analysis | 29/01/2025 | 11320033 |
2797888 | Certificate of Analysis | 28/01/2025 | 11320033 |
2645229 | Certificate of Analysis | 28/01/2025 | 11320033 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.