missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Leibovitz's L-15 Medium
GREENER_CHOICE

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue.

Brand:  Gibco™ 11415064

 View more versions of this product

Product Code. 15460554

  • £26.55 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Requires : Supplementation
This item is not returnable. View return policy

Description

Description

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.

This L-15 is modified as follows:

With: Galactose, Phenol Red, L-glutamine, Sodium Pyruvate

Without: Glucose, HEPES, Sodium Bicarbonate

The complete formulation is available.

L-15 contains no proteins, lipids, or growth factors. Therefore, F-10 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HEP-2 monkey kidney cells
Animal Origin
Liquid
Sterile-filtered
Glutamine, Phenol Red, Sodium Pyruvate, Galactose
cGMP-compliant under the ISO 13485 standard
500 mL
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
explants of embryonic and adult human tissue
1 X
Leibovitz's L-15 Medium
Sterile-filtered
No Glucose, No HEPES, No Sodium Bicarbonate
Gibco™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2766632 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number11415064
Lot Number2744459 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11415064
Lot Number2672052 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11415064
Lot Number2715490 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11415064
Lot Number2766630 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number11415064
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Leibovitz's L-15 Medium > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.