Learn More
Gilson™ PIPETMAN™ EasyPack™ Tips
GREENER_CHOICE
Ensure optimal performance of PIPETMAN pipettes across the full volume range with PIPETMAN™ EasyPack™ Tips.
Brand:  Gilson™ F161631
35.61 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
 View more versions of this product
Product Code. 10614353
  • £54.10 / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Optimized geometry and pure polypropylene material ensure low retention and naturally hydrophobic surface.
  • Manufactured in a cleanroom and hence no risk of contamination.
  • Full traceability from the mold to the bench guaranteed.
  • Volume graduations make it easy to visually check the volume aspirated.
  • Requires low tip-mounting force.
  • Easy identification with color-coded packaging.
  • Bulk packs are sealed with a zipper lock which can easily be opened for quick access and closed using the rapid-sealing strip to prevent contamination and loss of tips.
  • Each EasyPack box contains 1000 tips (5 bags of 200 tips or 1 bag of 1000; 4 bags of 50 tips or 3 bags of 334 tips).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1 to 10 μL
Non-sterile
Autoclavable
9.5 cm
Universal
Pipet Tips
Single-use
Pipetman EasyPack
Non-filtered
Clear
3.74 in.
Bagged
5 Bags of 200 Tips
Red
Yes
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ PIPETMAN™ EasyPack™ Tips > PIPETMAN™ EasyPack™ Tips; Volume: 0.1 to 10 μL; Quantity: 5 x 200 Tips/Box