missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ Glass Test Tubes for Photometer
Reliable, durable glass test tubes for all photometers.
Brand: Palintest™ PT595/5
This item is not returnable.
View return policy
Description
- X5 spare test tubes for all photometers
- Pack of 5
- For use with unique adaptive cell holder takes all reagent tubes 12mm to 20mm in diameter
- Coded to provide all the instrument needs for wavelength range and calibration set up and analysis; the user simply pushes in the tube
Specifications
Test tube | |
77 mm | |
Photometer; For Photometer | |
5 Pack | |
20 mm |
10 mL | |
Glass | |
Cap | |
Yes |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Palintest™ Glass Test Tubes for Photometer > Capacity: 10mL
Spot an opportunity for improvement?Share a Content Correction