missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HepatoZYME-SFM
GREENER_CHOICE

HepatoZYME-SFM

Brand:  Gibco™ 17705021

116.85 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


Product Code. 11570536

  • £125.00 / 500mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Hepatozyme-SFM is a serum-free medium for the long-term maintenance of hepatocyte phenotypic expression, including active and inducible forms of cytochromes P450 and active phase II enzymes.

Contains no phenol red.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C) and protect from light.
Serum-free
Liquid
Serum-free
500 mL
Hepatocyte Cells
Mammalian Cell Culture
Hepatocyte Serum Free Medium (SFM)
No Glutamine, No Phenol Red
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2517974 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number17705021
Lot Number2401429 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number17705021
Lot Number2517972 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number17705021
Lot Number2517976 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number17705021
Lot Number2517978 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number17705021
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ HepatoZYME-SFM > Quantity: 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.