Learn More
Hexahydro-4-methylphthalic anhydride, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -29°C |
Density | 1.1600g/mL |
Boiling Point | 120°C (1.0 Mbar) |
Flash Point | >110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4764 to 1.4784 |
Quantity | 2.5 kg |
Specific Gravity | 1.16 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0450668 | Certificate of Analysis | 02/05/2023 | 165395000,165390025 |
A0401246 | Certificate of Analysis | 15/12/2022 | 165395000,165390025 |
A0400500 | Certificate of Analysis | 15/12/2022 | 165395000,165390025 |
A0447202 | Certificate of Analysis | 16/11/2022 | 165395000,165390025 |
A0435383 | Certificate of Analysis | 18/10/2021 | 165395000,165390025 |
Safety and Handling


- Hexahydro-4-methylphthalic anhydride, mixture of cis and trans
- Danger
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H318-Causes serious eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.