Learn More
Nickel Electro™ Hinged Gable Lid
View samples better with a semi-transparent lid. Nickel Electro™ Hinged Gable Lid maximizes bath area while using its shape to drip condensation back into the bath.
Brand:  Nickel Electro™ GL1-14
 View more versions of this product
Product Code. 12768888
  • £123.00 / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket
Includes: dimForUseWithEquipment
Requires : ForUseWithApplication
Description
Description
  • Lightweight
  • Semi-transparent
  • Drips condensation back into bath
Transparent lid, hinged, high profile, handle
Specifications
Specifications
14 L Unstirred Baths
Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Nickel Electro™ Hinged Gable Lid > For Use With: For 14 Liter Unstirred Baths