Learn More
Epredia™ Histoplast Paraffin
Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.
Brand: Epredia™ 8331
82.36 GBP valid until 2025-04-30
Use promo code "24247" to get your promotional price.
Description
- Made of paraffin and plastic polymers
- Delivers excellent processing and yields thin, wrinkle-free sections
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).

Specifications
56°C | |
Paraffin IM |
Safety and Handling
- Histoplast IM
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P362-Take off contaminated clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.