missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ SE 400 Series Sturdier™ Air-Cooled Vertical Electrophoresis Systems
Economical units ideal for applications in which temperature control is not crucial
Brand: Hoefer™ SE400-15-1.5
Includes: Upper Buffer chamber, safety lid with electrodes, lower Buffer chamber and casting stand on leveling base with high voltage Leads, Glass plates, clamp assemblies, cams, slotted gasket for UBC, bubble level, laminated gasket for casting stand, spacer-mate-alignment template, wonder wedge-plate separation tool, gel seal
Requires : Comb and spacers must be ordered separately for Model SE 410 Basic.
This item is not returnable. View return policy
Description
- Units run one gel, or two with optional divider plate
- Built-in casting stand seals sandwiches leak-free without tape
- Rugged injection-molded construction for easy assembly and durability in high-use labs
- Plates, clamps, spacers and combs are interchangeable with SE 600 series units
Certifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE Marked.
Specifications
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE | |
Up to 80% | |
113°F | |
15°C to 30°C | |
0.15 cm | |
700 mL | |
7.08 in. | |
18 cm | |
6.29 in. | |
5.9 in. | |
500 V | |
60 mA |
18 x 16 cm | |
Upper Buffer chamber, safety lid with electrodes, lower Buffer chamber and casting stand on leveling base with high voltage Leads, Glass plates, clamp assemblies, cams, slotted gasket for UBC, bubble level, laminated gasket for casting stand, spacer-mate-alignment template, wonder wedge-plate separation tool, gel seal, comb, 15-well, 1.5 mm thick, spacers | |
45°C | |
0.05 in. | |
23.66 oz. | |
9.44 in. | |
24 cm | |
11.02 in. | |
28 cm | |
15 cm | |
20 w |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
HOE001 Hoefer Catalogue 2009 10
File size: 9.7 MB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hoefer™ SE 400 Series Sturdier™ Air-Cooled Vertical Electrophoresis Systems > SE 400; Gel Size: 18 x 16cm
Spot an opportunity for improvement?Share a Content Correction