Learn More
Fisherbrand™ Vertical Gel Tank
Incorporates a sealing system which is compatible with all major types of 80mm x 100mm and 100mm x 100mm precast gel. Fisherbrand™ Vertical Gel Tank's ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gel casting.
Brand: Fisherbrand™ VS10BI
Description
Incorporates a sealing system which is compatible with all major types of 80mm x 100mm and 100mm x 100mm precast gel. Gel casting and running utilise the same insert, no transfer of glass plates during gel casting is necessary. The insert contains sliding clamps allowing very rapid set up of both hand cast and precast gels. Ultra soft silicone seals and pressure bars which surround the glass plates guarantee leak proof gel casting. 2mm thick glass plates prevent breakage and have bonded spacers for convenience. Accessory electroblotting and tube gel modules are available which use the same outer tank and lid.- Injection molded construction, durable and leakproof
- Compatible with all 80mm x 100mm and 100mm x 100mm precast gels
- Low buffer volumes
- Run up to four gels
- Interchangable modules for IEF/2D electrophoresis and electroblotting in a universal tank For Mini SDS PAGE, native PAGE, gradient, second dimension and nucleic acid separations.
Specifications
80 mm x 100 mm and 100 mm x 100 mm PreCast Gels | |
Blotting Module Verti-gel Mini, 4 Cassettes, Fiber Pads | |
0.01 kg |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Fisherbrand and Fisher BioReagents Focus on electrophoresis