missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ HP35 Disposable Microtome Blades
Epredia™ HP35 microtome blades are coated for reliable and thin sectioning.
Brand: Epredia™ 3152735
This item is not returnable.
View return policy
Description
- High-profile
- Increased durability

Specifications
High Profile Blade | |
Designed for high-quality thin sectioning | |
None | |
0.01 in. |
High | |
Single-use | |
35° | |
0.31 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ HP35 Disposable Microtome Blades > Microtome blades; Non-coated
Spot an opportunity for improvement?Share a Content Correction