missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Immuno Tubes and Stoppers

Reproduce results time after time with these immuno tubes and stoppers, ideal for ELISA, solid phase RIA and liquid phase RIA assays.

Brand:  Thermo Scientific™ 341866

154.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10316141

  • £199.00 / Pack of 3600
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Polyethylene
11 mm
3600
11 mm Nunc-Immuno Tubes
Stopper
600
Not Applicable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number170000 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number341866
Lot Number165528 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number341866
Lot Number182339 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number341866
Lot Number180452 Certificate TypeCertificate of Compliance Date03/12/2024 Catalog Number341866
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Immuno Tubes and Stoppers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.