missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AccuPrime™ Taq DNA Polymerase, High Fidelity

Invitrogen AccuPrime Taq DNA Polymerase, High Fidelity, provides qualified reagents for the high-fidelity PCR amplification of nucleic acid templates.

Brand:  Invitrogen™ 12346086

 View more versions of this product

Product Code. 10125142

  • £475.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: AccuPrime™ Taq DNA Polymerase High Fidelity (40μL, at 5U/μL)) is supplied with: 50mM MgSO4 (1mL), 10X AccuPrime™ PCR Buffer I (1mL), 10X AccuPrime™ PCR Buffer II (1mL)

Buffer I and II differ in their concentration of thermostable AccuPrime™ protein. Their components are: 600mM Tris-SO4 (pH 8.9), 180mM (NH4)2SO4, 20mM MgSO4, 2mM dGTP, 2mM dATP, 2mM dTTP, 2mM dCTP, thermostable AccuPrime™ protein, 10% glycerol. The supplied PCR Buffer is a 10X concentrate and should be diluted for use. Buffer I is optimized for plasmids, cDNA, and λDNA. Buffer II is optimized for genomic DNA.
This item is not returnable. View return policy

Description

Description

Invitrogen AccuPrime Taq DNA Polymerase, High Fidelity, provides qualified reagents for the high-fidelity PCR amplification of nucleic acid templates. It includes an enzyme blend of Platinum Taq DNA Polymerase and a proofreading enzyme, plus AccuPrime accessory proteins for improved PCR fidelity, yield, and specificity over other hot-start DNA polymerases.

Features of AccuPrime Taq DNA Polymerase, High Fidelity

  • High specificity and yield for robust PCR amplification
  • 9-fold higher fidelity than Taq DNA polymerase alone
  • Mimimal optimization steps, even with non-optimized primer sets
  • Efficient amplification of targets over a broad size range up to 20 kb

How it works

Pyrococcus species GB-D polymerase is a proofreading enzyme that possesses a 3' to 5' exonuclease activity. Mixing this enzyme with Taq DNA Polymerase increases fidelity and allows amplification of simple and complex DNA templates over a large range of target sizes.

The Platinum antibody complexes with Taq DNA polymerase and inhibits activity at room temperature. Activity is restored after the initial denaturation step, providing an automatic hot start PCR.

The thermostable AccuPrime accessory proteins enhance specific primer-template hybridization during every cycle of PCR, preventing mispriming and enhancing PCR specificity and yield.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• AccuPrime Taq DNA Polymerase, High Fidelity (1 x 40 μL at 5 U/μL)
• 10X AccuPrime PCR Buffer I (1 x 1 mL)
• 10X AccuPrime PCR Buffer II (1 x 1 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -10°C to -30°C.
Low
Standard
200 rxns
Hot-start PCR, High-fidelity PCR
Built-In Hot Start
Mixed
20 kb or less
Primer-probe
AccuPrime Taq DNA Polymerase High Fidelity
DNA Polymerase
Dry Ice
9X
200 Reactions
Separate Components
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2460954 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12346086
Lot Number2460947 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12346086
Lot Number2536232 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12346086
Lot Number2500645 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12346086
Lot Number2611691 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number12346086
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ AccuPrime™ Taq DNA Polymerase, High Fidelity > 200 Reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.