Learn More
Jenway™ pH Temperature Electrode
Jenway™ pH Temperature Electrodes are used in conjunction with Jenway™ pH meters – 3510, 3520, 3540, 3340, 3345, and 4330.
Brand: Jenway™ 99960-43
Description
- Used in conjunction with Jenway™ pH meters – 3510, 3520, 3540, 3340, 3345, and 4330.
- Combination Type
- Refillable
- Reference Type: AgCl
- Isopotential: 0±20mV
Specifications
BNC | |
12 mm | |
Combination | |
120 mm | |
0 to 14 |
Flat end type | |
12 dia. x 120 mm L | |
Measurements of food extracts and agar plates, flat head makes it ideal for surface measurements | |
Epoxy | |
0°C to 80°C |
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.