Learn More
JZP 361, >99%, Tocris Bioscience™
Potent and selective reversible MAGL inhibitor
Brand:  Tocris Bioscience 5851/50
 View more versions of this product
Product Code. 30000842
  • £323.00 / 50mg
Explore more special offers
Description
Description
Potent and selective reversible MAGL inhibitor (IC50 = 46 nM). Exhibits 35-fold and 150-fold higher selectivity over human FAAH and ABHD6, respectively (IC50 values are 1.79 and 7.24 μM, respectively). Displays affinity for H1 receptors (pA2 = 6.81). Exhibits no activity at cannabinoid receptors at 10 μM.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
[4-(8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene)-1-piperidinyl](1H-1,2,4-
>99%
50 mg
405.88
1680193-80-9
C22H20ClN5O
Soluble to 100mM in DMSO and to 20mM in ethanol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
JZP 361, >99%, Tocris Bioscience™ > 50mg