Learn More
Brady™ LASERTAB™ Laser Printable Labels, B-747 Polyester
Designed for use with laser printers. Brady LASERTAB Laser Printable Labels are made of B-747 polyester.
£178.00 - £615.00
Specifications
Adhesive Type | Acrylic, Permanent |
---|---|
Finish | Matte |
Material | Polyester (B-747) |
Product Type | Laser Printable Label |
Thickness (English) | 0.0037 in. |
Product Code | Brand | Size (Metric) | Size (English) | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size (Metric) | Size (English) | Color | Price | Quantity & Availability | |||
11573584
|
Brady™
029730 |
47.6 x 9.52 mm (W x H) | 1.875 x 0.375 in. (W x H) | White |
£349.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11583584
|
Brady™
029733 |
76.2 x 9.52 mm (W x H) | 3 x 0.375 in. (W x H) | Yellow |
£253.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11503594
|
Brady™
029736 |
47.6 x 21.1 mm (W x H) | 1.875 x 0.833 in. (W x H) | White |
£312.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11503644
|
Brady™
029794 |
47.6 x 21.1 mm (W x H) | 1.875 x 0.833 in. (W x H) | Yellow |
£389.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11513594
|
Brady™
029738 |
47.6 x 33.7 mm (W x H) | 1.875 x 1.33 in. (W x H) | White |
£320.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11583564
|
Brady™
029700 |
7.62 x 6.35 mm (W x H) | 0.3 x 0.25 in. (W x H) | White |
£247.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11513644
|
Brady™
029795 |
63.5 x 25.4 mm (W x H) | 2.5 x 1 in. (W x H) | Yellow |
£283.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11533614
|
Brady™
029762 |
12.7 x 11.09 mm (W x H) | 0.5 x 0.437 in. (W x H) | Yellow |
£234.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11543614
|
Brady™
029763 |
6.35 x 6.35 mm (W x H) | 0.25 x 0.25 in. (W x H) | White |
£247.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11503574
|
Brady™
029703 |
9.52 x 9.52 mm (W x H) | 0.375 x 0.375 in. (W x H) | White |
£245.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11593614
|
Brady™
029768 |
215.9 x 279.4 mm (W x H) | 8.5 x 11 in. (W x H) | White |
£178.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11523644
|
Brady™
029796 |
215.9 x 279.4 mm (W x H) | 8.5 x 11 in. (W x H) | Yellow |
£195.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11523624
|
Brady™
029771 |
16.5 x 5.08 mm (W x H) | 0.65 x 0.2 in. (W x H) | White |
£247.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11523634
|
Brady™
029785 |
16.5 x 5.08 mm (W x H) | 0.65 x 0.2 in. (W x H) | Yellow |
£271.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11523574
|
Brady™
029706 |
16.5 x 6.35 mm (W x H) | 0.65 x 0.25 in. (W x H) | White |
£247.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11543574
|
Brady™
029709 |
20.3 x 6.35 mm (W x H) | 0.8 x 0.25 in. (W x H) | White |
£340.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11563574
|
Brady™
029712 |
20.3 x 12.7 mm (W x H) | 0.8 x 0.5 in. (W x H) | White |
£520.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11593574
|
Brady™
029718 |
25.4 x 12.7 mm (W x H) | 1 x 0.5 in. (W x H) | Yellow |
£560.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
11553634
|
Brady™
029789 |
25.4 x 12.7 mm (W x H) | 1 x 0.5 in. (W x H) | White |
£615.00
1 pack |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11533584
|
Brady™
029724 |
38.1 x 6.35 mm (W x H) | 1.5 x 0.25 in. (W x H) | White |
£353.00
1 pack |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
Description
- Blank, printable polyester labels for Laser Printers
- CSA Accepted material when printed with designated laser printer toners
Specifications
Acrylic, Permanent | |
Polyester (B-747) | |
0.0037 in. |
Matte | |
Laser Printable Label |