missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Leibovitz's L-15 Medium, powder

Leibovitz's L-15 Medium, powder

Brand:  Gibco™ 41300021

Product Code. 10237252

  • £9.13 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Leibovitz's L-15 Medium supports the growth of HEP-2 monkey kidney cells and primary explants of embryonic and adult human tissue. L-15 is buffered by phosphates and free base amino acids instead of sodium bicarbonate. This medium is designed for supporting cell growth in environments without CO2 equilibration.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ L-15 is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ L-15 product made in our Grand Island facility (41300-039). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

L-15 contains no proteins, lipids, or growth factors. Therefore, L-15 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS).

Powder forms of Gibco™ L-15 cell culture medium require pH adjustment and filtration at the time of preparation (see protocol for details).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 13485 standards, FDA
Leibovitz's L-15 Medium, 1L, Powder, 7.7 to 8.1 pH, With Galactose, Phenol Red, L-glutamine, Sodium Pyruvate, 36 Months Shelf Life, 290 to 330mOsm/kg Osmolality, For Mammalian Cell Culture
Leibovitz's L-15 Medium
No Glucose, No HEPES, No Sodium Bicarbonate
7.7 to 8.1
2 to 8°C
Powder
Glutamine, Galactose, Phenol Red, Sodium Pyruvate
Mammalian cell culture
1 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Leibovitz's L-15 Medium, powder > 1 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.