Learn More
Lovibond™ Total Chlorine Powder Packs™
Premeasured unit dose DPD reagents for water quality testing
Brand:  Lovibond™ 530120
 View more versions of this product
Product Code. 10402345
  • £22.10 / Pack of 100
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Premeasured unit dose DPD reagents for water quality testing
  • For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed foil packets eliminate risk of cross-contamination
  • Available for 5, 10, and 25mL sample sizes
  • Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
Specifications
US EPA
Powder
SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
0 to 2 mg/L
0.33 oz.
10 mL
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
100 Pack
DPD Total Chlorine Reagent
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lovibond™ Total Chlorine Powder Packs™ > Total Chlorine; 10mL; Pack of 100