Learn More
Axygen™ Mini Tube System
Polypropylene tube system for compound library storage
£38.21 - £565.78
Specifications
Color | Clear |
---|---|
For Use With (Application) | Ensures positive sample identification |
Product Code | Brand | Sterility | Capacity (Metric) | Packaging | Bottom Shape | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Capacity (Metric) | Packaging | Bottom Shape | Price | Quantity & Availability | ||||||
12529137
|
Axygen™
MTS-8CP-C |
Non-sterile | 8 Cap Strip | 120 Strips/PK., 5 Packs/Cs. | Strip |
£102.20
Pack of 600 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
11326324
|
Axygen™
MTS-11-8-C-R-S |
Sterile | 1.1 mL | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round | N/A | N/A | N/A | |||||
12716829
|
Axygen™
MTS-11-8-C-R-S |
Sterile | 1.1 mL | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round |
£424.36
Pack of 600 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
12598007
|
Axygen™
MTS-12CP-C |
Non-sterile | 12 Cap Strip | 80 Strips/PK., 5 Packs/Cs. | Strip |
£98.27
Pack of 400 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
11386314
|
Axygen™
MTS-11-12-C-R |
Non-sterile | 1.1 mL | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round | N/A | N/A | N/A | |||||
12786819
|
Axygen™
MTS-11-12-C-R |
Non-sterile | 1.1 mL | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round |
£353.50
Pack of 400 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
10313030
|
Axygen™
MTS-11-12-C-R-S |
Sterile | 1.1 mL | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round |
£424.36
Pack of 400 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
12549207
|
Axygen™
MTS-11-12-C-R-S |
Sterile | 1.1 mL | 8 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round | N/A | N/A | N/A | |||||
12746829
|
Axygen™
MTS-11-C-R-S |
Sterile | 1.1 mL | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round |
£424.36
Pack of 4800 |
Estimated Shipment: 23-06-2025
Log in to see stock. |
||||||
11336324
|
Axygen™
MTS-11-C-R-S |
Sterile | 1.1 mL | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round | N/A | N/A | N/A | |||||
12539137
|
Axygen™
MTS-8CP-C-S |
Sterile | 8 Cap Strip | 120 Strips/PK., 5 Packs/Cs. | Strip |
£138.73
Pack of 600 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
13117664
|
Axygen™
MTS-06-C-R |
Non-sterile | 0.65 mL | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round |
£458.17
Pack of 4800 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
11356314
|
Axygen™
MTS-06-C-R |
Non-sterile | 0.65 mL | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round | N/A | N/A | N/A | |||||
12508017
|
Axygen™
MTS-12CP-C-S |
Sterile | 12 Cap Strip | 80 Strips/PK., 5 Packs/Cs. | Strip |
£138.73
Pack of 400 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
12776819
|
Axygen™
MTS-06-C-R-S |
Sterile | 0.65 mL | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round |
£565.78
Pack of 4800 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
11366314
|
Axygen™
MTS-06-C-R-S |
Sterile | 0.65 mL | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round | N/A | N/A | N/A | |||||
12588007
|
Axygen™
MTS-11-12-C |
Non-sterile | 1.1 mL | 80 Strips/Bag; 5 Bags/Cs. | Round |
£155.09
Pack of 400 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
13121794
|
Axygen™
MTS-11-8-C-R |
Non-sterile | 1.1 mL | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round |
£353.50
Pack of 600 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
11316324
|
Axygen™
MTS-11-8-C-R |
Non-sterile | 1.1 mL | 12 Strips/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round | N/A | N/A | N/A | |||||
12736829
|
Axygen™
MTS-11-C-R |
Non-sterile | 1.1 mL | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round |
£391.00
Pack of 4800 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
12559207
|
Axygen™
MTS-11-C-R |
Non-sterile | 1.1 mL | 96/Rack, 10 Racks/Pk., 5 Packs/Cs. | Round | N/A | N/A | N/A | |||||
12519137
|
Axygen™
MTS-11-8-C |
Non-sterile | 1.1 mL | 120 Strips/Bag; 5 Bags/Cs. | Round |
£155.09
Pack of 600 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
12531288
|
Axygen™
MTS-06-C |
Non-sterile | 0.65 mL | 960 Tubes/Bag, 5 Bags/Cs. | Round |
£206.73
Pack of 4800 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
12541288
|
Axygen™
MTS-11-C |
Non-sterile | 1.1 mL | 960 Tubes/Bag, 5 Bags/Cs. | Round |
£155.09
Pack of 4800 |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
Description
Axygen™ Mini Tube Systems are available in 0.65 mL single mini tube or 1.1 mL single mini tube. They are also available in 8-cap or 12-cap strip format. Mini Tube System racks are compatible with Beckman Coulter, Tecan, and other automation workstations.
- Rack features a one-way lid design and alpha-numeric grid
- Molded-in condensation rings on the lid minimize cross contamination
Specifications
Clear | |
Ensures positive sample identification |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.