Learn More
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™
Versatile check valves can be installed in any position
Brand:  Cole-Parmer 50115-21
 View more versions of this product
Product Code. 30053078
  • £4.10 / Each
Estimated Shipment: 18-11-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Prevent backflow to protect your equipment. Work with both liquids and gases. Use for pressure or vacuum applications. 0.18 psi breaking pressure
Specifications
Specifications
0.1875 in.
3/16 in. hose barb
Viton
0.12
Barbed
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer Miniature Check Valves, Hose Barb, Polypropylene and Viton™ > 3/16 in. hose barb