Learn More
MP Biomedicals™ ETBR Greenbag Disposal Kit
ETBR Greenbag Disposal Kit for removing Ethidium Bromide from solutions.
Brand: MP Biomedicals™ 112350200
361.40 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Each kit can remove 500 mg of Ethidium Bromide
- Each bag contains a standard amount of activated material that will safely and easily concentrate Ethidium Bromide from large liquid quantities
- The bags containing the biohazardous material can then be disposed of in a proper container
- Applications include Agarose Gel Electrophoresis, DNA and RNA purification
- Store at room temperature 15°C to 30°C
- 50 bags/kit

Specifications
ETBr Green Bag Disposal Kit | |
Single-use | |
50 Bags/Pk. |
Store at 15°C to 30°C | |
Agarose gel electrophoresis, DNA, RNA purification |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
5Hours
2Minutes
37Seconds
17
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.