Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ The Flip™ Safety Pipette Fillers are manufactured from natural rubber.
Brand: Deutsch Neumann™ 4710001
Description
- Only two operating points
- Evacuate via the new automatic valve
- Available in a range of colors
- One-handed design is the simplest safety pipette filler available
- Ability to clean inside of bulb by removing patented valve and rinsing out
Specifications
Red | |
Natural Rubber | |
Safety Pipette Filler Flip |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 13
Product Content Correction
Product Title
Deutsch Neumann™ The Flip™ Safety Pipette Fillers > Red
Spot an opportunity for improvement?Share a Content Correction