missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELISA Stop Solution

A ready-to-use solution intended for use as a stop solution in ELISA applications that use horseradish peroxidase and TMB substrate solution.

Brand:  Invitrogen™ SS03

70.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12043609

  • £74.75 / 25mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

ELISA Stop Solution is a ready-to-use solution intended for use as a stop solution in ELISA applications that use horseradish peroxidase and TMB substrate solution. As TMB substrate solution reacts with horseradish peroxidase, it produces a pronounced blue color. At the end of TMB incubation, a volume of ELISA Stop Solution equal to the volume of TMB substrate solution is added to the wells to terminate the horseradish peroxidase reaction. Addition of Stop Solution results in the rapid formation of a yellow end product. The optical density of this yellow end product is measured at 450 nm. The yellow end product is stable for sixty minutes after addition of Stop Solution and so optical density should be measured within that time.

For a complete kit of ELISA buffers, see ELISA Buffer Kit.

View all available buffers and reagents for ELISA applications.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

ELISA Stop Solution
Liquid
Store at 2°C to 8°C
25 mL
HRP, TMB
Ambient
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number441188-000 Certificate TypeCertificate of Analysis Date13/03/2025 Catalog Number
Lot Number436995-000 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog Number
Lot Number436995-000 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberSS03
Lot Number431195-000 Certificate TypeCertificate of Analysis Date14/11/2024 Catalog NumberSS03
Lot Number428490-000 Certificate TypeCertificate of Analysis Date24/10/2024 Catalog NumberSS03
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ELISA Stop Solution > 25 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.