Learn More
Invitrogen™ XCell4 SureLock™ Midi-Cell
Easily set up in just three simple steps
Brand: Invitrogen™ WR0100
Includes: XCell4 Lower Buffer Chamber; XCell4 Lid with Cables; 2 XCell4 Buffer Cores; XCell4 Buffer Dam; XCell4 Displacement Dam; XCell4 Surelock Assembly; Gel Knife; Set of 2 Power Supply adapters (Zoom Adapters)
Description
The XCell4 SureLock Midi-Cell allows you to quickly and easily perform simultaneous vertical electrophoresis of 1–4 midi-gels without leaking, resulting in consistent performance. The XCell4 SureLock Midi-Cell is built with tough plastic material to boost durability and features a specially designed lid to enhance user safety.
- Uses patented technology to make electrophoresis easier and more reliable
- Designed to dissipate heat effectively and evenly, and ensure high-resolution results when using Novex midi gels (8.7 x 13.3cm)
Flexible, Efficient, and Easy to Use
- Simple assembly and operation—a single SureLock actuator seals all gels while allowing for zero force insertion and easy positioning of the gels for sample loading
- Runs up to four Novex midi-gels at once, while having the same approximate footprint as the Criterion™ Cell (Bio-Rad), which runs a maximum of only two gels
- No need for external cooling accessories–requires 25% less running buffer per gel than the Criterion Cell (340mL vs. 460mL)
- The XCell4 SureLock Midi-Cell consists of a Lower Buffer Chamber, two Buffer Cores, a lid with cables, a Displacement Dam, and a Buffer Dam
- Electrophoresis of proteins on midi-gels in the XCell4 SureLock Midi-Cell is performed after assembling each Buffer Core with two midi-gels or a Buffer Core with a midi-gel and a Buffer Dam
- The Buffer Core/gel assembly is inserted into the Lower Buffer Chamber using a positive locking mechanism that seals the Buffer Core/gel assembly firmly into position and ensures leak-free electrophoresis
- The Buffer Core/gel assembly creates separate upper buffer chambers for each gel and a common lower buffer chamber shared by all gels
- The lid is placed onto the Midi-Cell and fits on the Midi-Cell in only one direction
- The XCell4 SureLock Midi-Cell holds gels firmly with a simple gel tension wedge; when the lever on the gel tension wedge is pushed forward into the locked position, an even horizontal force is generated, which seals the gel/buffer core assembly firmly into position in the lower buffer chamber
- The positive locking action of the gel tension wedge ensures a trouble-free, leak-free gel run every time
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature

Specifications
The components of the midi-cell include: • 1 XCell4 Lower Buffer Chamber • 1 XCell4 Lid with Cables • 2 XCell4 Buffer Cores • 1 XCell4 Buffer Dam • 1 XCell4 Displacement Dam • 1 XCell4 Surelock Assembly • 1 Gel Knife • 1 Set of 2 Power Supply adapters (Zoom Adapters). Operating temperature is 4°C to 40°C. The XCell4 Surelock Midi-Cell Runner is guaranteed for 1 year when properly stored. |
|
Novex™ Midi Gels | |
XCell SureLock | |
1 Each | |
600 VDC |
XCell4 SureLock™ Midi | |
Midi (8 x 13 cm) | |
Midi-Cell Midi-Gel Electrophoresis System | |
Room Temperature |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.