missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Kit

Clean up PCR reactions and gel extractions with a two-in-one approach. Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Kit is optimized for recovery and elution volume when purifying DNA fragments via both applications.

Brand:  Macherey-Nagel™ 740609.250

392.19 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 11992242

  • £725.00 / 250 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

NucleoSpin™ Gel and PCR Clean-up is designed for fast purification of PCR products, such as DNA from enzymatic reactions, as well as the extraction of DNA fragments from TAE or TBE agarose gels.

  • Two applications in one kit—One buffer to process both sample types, with only one loading step
  • High recovery for fragments down to 50 bp—Average recovery from 60–90%
  • Minimized elution volume of 15μL—Highly concentrated DNA
  • Binding buffer with pH indicator—Ensures optimal binding conditions
  • Separate buffers for single stranded DNA and RNA or for SDS-containing samples available separately
  • Suitable for all kinds of agarose gels and a variety of buffer systems (e.g. TAE, TBE)
  • Complete removal of contaminations: nucleotides, primers, enzymes, mineral oil, PCR additives, detergents, dyes
  • Applications: purification of PCR products, extraction of DNA from agarose or polyacrylamide gels, and downstream applications (cloning, sequencing, PCR, restriction analysis)

Kit contents: NucleoSpin™ Gel and PCR Clean-up columns, collection tubes (2mL), binding buffer with pH indicator, wash buffer and elution buffer

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.

Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ NucleoSpin™ Gel and PCR Clean-up Kit > No. of Reactions: 250

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.