Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils

Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Brand: Thermo Scientific™ 5701
45.17 GBP valid until 2025-06-30
Use promo code "25075" to get your promotional price.
Description
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
Protect microplate well contents with the Thermo Scientific™ Mylar™ Plate Sealer for Microtiter™ Plates, which assures a tight seal. Designed for use when sealing microtiter plates.
Specifications
Clear | |
SEALER, MYLAR, 100/CS | |
Non-sterile | |
96 | |
100 | |
Short-term storage, ELISA, minimizes evaporation and cross contamination | |
Adhesive Seal | |
Acrylic | |
No | |
All well shapes | |
General |
Life Science Reagent Resistant | |
Polyester | |
Not Autoclavable | |
100 | |
83 x 124 mm | |
General | |
Nunc | |
No | |
-40°C to +120°C | |
All Microplates | |
Yes |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Thermo Scientific Matrix
Exclusive Discounts on Storage Solutions: Storage Tubes, Equipment and Accessories
Upgrade your lab with top-quality storage consumables and equipment at unbeatable prices.Promo Code: 25075Expires: 2025-06-30
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.