Learn More
WTW™ OxiTop™-i Respirometric BOD Measuring System
The WTW™ OxiTop™-i Respirometric BOD Measuring System is designed for self-check BOD measurement with undiluted samples (according to DIN EN 1899 -2) at measuring ranges up to 4000 mg/L.
Brand:  WTW™ 208260
2665.91 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16219020
  • £2820.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • OxiTop™ is a respirometric vessel-top measuring system for BOD self-check measurement with undiluted samples (DIN EN 1899 -2)
  • Measuring ranges up to 4000 mg/L BOD possible
  • No need to convert data - direct input of sample volume
  • Direct display of the BOD value on the measuring head screen
  • Menu-operated, with backlit graphic LCD, control LED and operating buttons for direct BOD readings, volume pre-selection, measurement duration between 1 and 7 days selectable, display of measuring curve, display of current readings
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OxiTop™-i IS 6
Backlit Graphic LCD
Measuring ranges up to 4000 mg/L BOD possible
Six OxiTop™-i measuring heads, universal plug-in power supply 100-240 VAC, 50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
WTW™ OxiTop™-i Respirometric BOD Measuring System > Type: OxiTop™-i IS 6