missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brain Heart Infusion Broth (Dehydrated)

Cultivate streptococci, Neisseria and other fastidious organisms with this highly nutritious medium.

Brand:  Thermo Scientific™ CM1135B

116.61 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 11469668

  • £221.00 / 500g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Cultivate streptococci, Neisseria and other fastidious organisms with Thermo Scientific™ Oxoid™ Brain Infusion Broth (Dehydrated). The medium is highly nutritious and well buffered to support the growth of wide variety of organisms. It is recommended for blood culture work for the isolation and cultivation of pathogenic fungi.

Cultivate a wide variety of organisms with Brain Infusion Broth:

  • Use as a growth and suspension medium for staphylococci when testing coagulase production.
  • Add 10% defibrillated blood for isolation and cultivation of pathogenic fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Brain Heart Infusion Broth
Dehydrated Culture Media
For 13.5L medium
Powder
Each
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6224624 Certificate TypeCertificate of Analysis Date04/04/2025 Catalog NumberCM1135B
Lot Number6236076 Certificate TypeCertificate of Analysis Date25/03/2025 Catalog NumberCM1135B
Lot Number6235486 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberCM1135B
Lot Number6206070 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberCM1135B
Lot Number6206069 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberCM1135B
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Brain Heart Infusion Broth (Dehydrated) > 500g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.