Learn More
Cytiva PEEK Union
Connect piping of varying sizes. Cytiva Life Sciences™ PEEK Unions are an important accessory for various scientific applications.
Brand:  Cytiva 18-1142-08
 View more versions of this product
Product Code. 12534317
  • £43.68 / Pack of 8
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Packsize 5
  • When ordered separately the connectors are black.
  • Made of polyetheretherketone (PEEK)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Union
5/16 in. Female to 1/16 in. Male
PEEK
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    7
    Hours
    19
    Minutes
    3
    Seconds
    32
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127
    Expires2025-05-31
    Days
    38
    Hours
    19
    Minutes
    3
    Seconds
    32
Product Content Correction
Product Title
Cytiva PEEK Union > 5/16 in. Female to 1/16 in. Male Connection