Documents
Scientific Resources
Application Notes
- 1
- 2
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Brand: Thermo Scientific™ 436009
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (CovaLink™ and Immobilizer Amino)
Standard | |
Colorimetric | |
30 Cases | |
Clear | |
Polystyrene | |
384 | |
Non-sterile | |
120 μL | |
30 | |
Unspecified | |
Peptides |
Without Lid | |
16 x 24 | |
Covalent | |
Solid plate 384-Well | |
120 μL | |
50 μL | |
Amine | |
Flat Bottom (F-well) | |
5 | |
30/Cs. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.