missing translation for 'onlineSavingsMsg'
Learn More
Witeg™ Polypropylene Pouring Ring Product Code.: 11815145
In Stock
Add to basket

Witeg™ Polypropylene Pouring Ring

Product Code. 11815145
Polypropylene
Click to view available options
Material:
Polypropylene

Product Code. 11815145

Brand: Witeg™ 5 526 166

In Stock
Edge
Add to basket
Add to basket

Autoclavable. Witeg™ Polypropylene Pouring Ring are compatible with GL 45 Screw Thread Laboratory Bottles.

  • Polypropylene
  • Yellow
  • GL 45

Specifications

Color Yellow
Material Polypropylene
No. per Pack 10Pack
Quantity 10Pack
Autoclavable Autoclavable
Product Type Pouring Ring
For Use With (Equipment) For GL 45 thread
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Witeg™ Polypropylene Pouring Ring >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.