missing translation for 'onlineSavingsMsg'
Learn More
Pyrazine-2-carboxylic acid, 99%, Thermo Scientific Chemicals Product Code.: 11470993

Pyrazine-2-carboxylic acid, 99%, Thermo Scientific Chemicals

Product Code. 11470993
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11470993

Brand: Thermo Scientific Alfa Aesar A13363.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-97-5
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.099
MDL Number MFCD00006130
InChI Key NIPZZXUFJPQHNH-UHFFFAOYSA-N
Synonym 2-pyrazinecarboxylic acid, pyrazinoic acid, pyrazinecarboxylic acid, pyrazinic acid, 2-carboxypyrazine, paradiazinecarboxylic acid, pyrazinemonocarboxylic acid, 2-pyrazinoic acid, 2-pyrazine carboxylic acid, piazinecarboxylic acidShow MoreShow Less
PubChem CID 1047
ChEBI CHEBI:71311
IUPAC Name pyrazine-2-carboxylic acid
SMILES C1=CN=C(C=N1)C(=O)O

Specifications

Melting Point ∼225°C (decomposition)
Quantity 25 g
Beilstein 112305
Merck Index 14,7959
Formula Weight 124.1
Percent Purity 99%
Chemical Name or Material Pyrazine-2-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10237893 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13363.14,A13363.22
Lot Number10237520 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13363.14,A13363.22
Lot Number10142950 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13363.14,A13363.22
Lot NumberJ7134A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13363.14,A13363.22
Lot Number10235782 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13363.14,A13363.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrazine-2-carboxylic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.