Learn More
Thermo Scientific™ Sample Storage Starter Packs
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs
Brand: Thermo Scientific™ HHD-HSX-WP1D-2
Description
Introduce automation into your workflow with Thermo Scientific™ sample storage starter packs. Choose from five different starter pack options: BASIC, ADVANCED, PREMIUM, PREMIUM 0.5 mL and PREMIUM 1.mL. All Starter Packs include Matrix™ ScrewTop Tri-coded Tubes with an 8-Channel Screw Cap Decapper. Advanced and Premium Starter Packs also include a barcode reader.
Engineered with the value of your samples in mind:
- Choose from five different starter pack options.
- Eliminate the need to reformat non-2D cryotube samples.
- Secure sample tracking with 2D, 1D, and human readable codes.
- Cap/decap a full rack of tubes in less than 1 minute.
- Scan 96 tubes in as little as 1 second with advanced and premium starter packs.
- Help ensure quality with ISO-7 class cleanroom– manufactured tubes with sterility assurance.
Specifications
Premium Starter Pack | |
Tubes, Decapper, HSX Reader |
Product Suggestions
Customers who viewed this item also viewed